Anti-FCRL5 Mouse Monoclonal Antibody [clone: 307307]

Supplier: R & D Systems
MAB20871
220019-582EA 639.2 USD
220019-582
Anti-FCRL5 Mouse Monoclonal Antibody [clone: 307307]
Antibodies
Type: Primary
Antigen Symbol: FCRL5
Antigen Name:
Antigen Synonyms: FCRL3
Clonality: Monoclonal
Clone: 307307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR