Anti-FCRL5 Mouse Monoclonal Antibody [clone: 307314]

Supplier: R & D Systems
MAB2087
220019-581EA 600.15 USD
220019-581
Anti-FCRL5 Mouse Monoclonal Antibody [clone: 307314]
Antibodies
Type: Primary
Antigen Symbol: FCRL5
Antigen Name: Fc receptor-like 5
Antigen Synonyms: BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820
Clonality: Monoclonal
Clone: 307314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR