Anti-GAD1 Mouse Monoclonal Antibody [clone: 252910]

Supplier: R & D Systems
MAB2086
220019-580EA 577.02 USD
220019-580
Anti-GAD1 Mouse Monoclonal Antibody [clone: 252910]
Antibodies
Type: Primary
Antigen Symbol: GAD1
Antigen Name: glutamate decarboxylase 1 (brain, 67kDa)
Antigen Synonyms: CPSQ1, GAD, SCP
Clonality: Monoclonal
Clone: 252910
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR