Anti-AMIGO2 Mouse Monoclonal Antibody [clone: 273617]

Supplier: R & D Systems
MAB2080
220019-577EA 577.02 USD
220019-577
Anti-AMIGO2 Mouse Monoclonal Antibody [clone: 273617]
Antibodies
Type: Primary
Antigen: AMIGO2
Clonality: Monoclonal
Clone: 273617
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR