Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 239421]

Supplier: R & D Systems
MAB2079
220019-575EA 594.37 USD
220019-575
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 239421]
Antibodies
Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 239421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR