Anti-LILRB2 Mouse Monoclonal Antibody [clone: 287219]

Supplier: R & D Systems
MAB2078
220019-574EA 655.11 USD
220019-574
Anti-LILRB2 Mouse Monoclonal Antibody [clone: 287219]
Antibodies
Type: Primary
Antigen: LILRB2
Clonality: Monoclonal
Clone: 287219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR