Anti-JNK1 / JNK2 Mouse Monoclonal Antibody [clone: 252323]

Supplier: R & D Systems
MAB2076
220019-573EA 594.37 USD
220019-573
Anti-JNK1 / JNK2 Mouse Monoclonal Antibody [clone: 252323]
Antibodies
Type: Primary
Antigen: JNK1 / JNK2
Clonality: Monoclonal
Clone: 252323
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR