Anti-CTSF Mouse Monoclonal Antibody [clone: 237208]

Supplier: R & D Systems
MAB2075
220019-572EA 772.25 USD
220019-572
Anti-CTSF Mouse Monoclonal Antibody [clone: 237208]
Antibodies
Type: Primary
Antigen: CTSF
Clonality: Monoclonal
Clone: 237208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR