Anti-FCGR1 Rat Monoclonal Antibody [clone: 290322]

Supplier: R & D Systems
MAB20741
220019-571EA 633.42 USD
220019-571
Anti-FCGR1 Rat Monoclonal Antibody [clone: 290322]
Antibodies
Type: Primary
Antigen Symbol: FCGR1
Antigen Name: Fc receptor, IgG, high affinity I
Antigen Synonyms: AI323638, AV092959, CD64, FcgammaRI, IGGHAFC
Clonality: Monoclonal
Clone: 290322
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR