Anti-WFIKKN2 Mouse Monoclonal Antibody [clone: 288504]

Supplier: R & D Systems
MAB20702
220019-569EA 763.57 USD
220019-569
Anti-WFIKKN2 Mouse Monoclonal Antibody [clone: 288504]
Antibodies
Type: Primary
Antigen: WFIKKN2
Clonality: Monoclonal
Clone: 288504
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR