Anti-WFIKKN2 Mouse Monoclonal Antibody [clone: 288518]

Supplier: R & D Systems
MAB20701
220019-568EA 561.11 USD
220019-568
Anti-WFIKKN2 Mouse Monoclonal Antibody [clone: 288518]
Antibodies
Type: Primary
Antigen: WFIKKN2
Clonality: Monoclonal
Clone: 288518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR