Anti-WFIKKN2 Mouse Monoclonal Antibody [clone: 288513]

Supplier: R & D Systems
MAB2070
220019-567EA 577.02 USD
220019-567
Anti-WFIKKN2 Mouse Monoclonal Antibody [clone: 288513]
Antibodies
Type: Primary
Antigen: WFIKKN2
Clonality: Monoclonal
Clone: 288513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR