Anti-GREM2 Rat Monoclonal Antibody [clone: 377407]

Supplier: R & D Systems
MAB2069
220019-565EA 561.11 USD
220019-565
Anti-GREM2 Rat Monoclonal Antibody [clone: 377407]
Antibodies
Type: Primary
Antigen Symbol: GREM2
Antigen Name: gremlin 2 homolog, cysteine knot superfamily (Xenopus laevis)
Antigen Synonyms: Gremlin2, Prdc
Clonality: Monoclonal
Clone: 377407
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR