Anti-SEMA7A Rat Monoclonal Antibody [clone: 238906]

Supplier: R & D Systems
MAB2068
220019-563EA 763.57 USD
220019-563
Anti-SEMA7A Rat Monoclonal Antibody [clone: 238906]
Antibodies
Type: Primary
Antigen: SEMA7A
Clonality: Monoclonal
Clone: 238906
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR