Anti-LAMB2 Mouse Monoclonal Antibody [clone: C4]

Supplier: R & D Systems
MAB2066
220019-561EA 642.09 USD
220019-561
Anti-LAMB2 Mouse Monoclonal Antibody [clone: C4]
Antibodies
Type: Primary
Antigen: LAMB2
Clonality: Monoclonal
Clone: C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR