Anti-S100 A9 Rat Monoclonal Antibody [clone: 372510]

Supplier: R & D Systems
MAB2065
220019-560EA 594.37 USD
220019-560
Anti-S100 A9 Rat Monoclonal Antibody [clone: 372510]
Antibodies
Simple Western

Type: Primary
Antigen: S100 A9
Clonality: Monoclonal
Clone: 372510
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR