Anti-C1S Mouse Monoclonal Antibody [clone: 306904]

Supplier: R & D Systems
MAB2060
220019-557EA 594.37 USD
220019-557
Anti-C1S Mouse Monoclonal Antibody [clone: 306904]
Antibodies
Type: Primary
Antigen: C1S
Clonality: Monoclonal
Clone: 306904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR