Anti-PRSS22 Rat Monoclonal Antibody [clone: 246702]

Supplier: R & D Systems
MAB20592
220019-555EA 763.57 USD
220019-555
Anti-PRSS22 Rat Monoclonal Antibody [clone: 246702]
Antibodies
Type: Primary
Antigen: PRSS22
Clonality: Monoclonal
Clone: 246702
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR