Anti-IL17F Rat Monoclonal Antibody [clone: 316016]

Supplier: R & D Systems
MAB2057
220019-552EA 594.37 USD
220019-552
Anti-IL17F Rat Monoclonal Antibody [clone: 316016]
Antibodies
Type: Primary
Antigen: IL17F
Clonality: Monoclonal
Clone: 316016
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR