Anti-RSK Mouse Monoclonal Antibody [clone: 792921]

Supplier: R & D Systems
MAB2056
102885-424EA 633.42 USD
102885-424
Anti-RSK Mouse Monoclonal Antibody [clone: 792921]
Antibodies
Simple Western

Type: Primary
Antigen: RSK
Clonality: Monoclonal
Clone: 792921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR