Anti-AKT (pan specific) Mouse Monoclonal Antibody [clone: 28146]

Supplier: R & D Systems
MAB2055
220019-551EA 655.11 USD
220019-551
Anti-AKT (pan specific) Mouse Monoclonal Antibody [clone: 28146]
Antibodies
Simple Western

Type: Primary
Antigen: AKT (pan specific)
Clonality: Monoclonal
Clone: 28146
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR