Anti-MIA Mouse Monoclonal Antibody [clone: 294203]

Supplier: R & D Systems
MAB20501
220019-550EA 577.02 USD
220019-550
Anti-MIA Mouse Monoclonal Antibody [clone: 294203]
Antibodies
Type: Primary
Antigen: MIA
Clonality: Monoclonal
Clone: 294203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR