Anti-FCRLA Mouse Monoclonal Antibody [clone: 282415]

Supplier: R & D Systems
MAB2049
220019-547EA 577.02 USD
220019-547
Anti-FCRLA Mouse Monoclonal Antibody [clone: 282415]
Antibodies
Type: Primary
Antigen Symbol: FCRL1
Antigen Name: Fc receptor-like 1
Antigen Synonyms: CD307a, FCRH1, IFGP1, IRTA5
Clonality: Monoclonal
Clone: 282415
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR