Anti-LY75 Mouse Monoclonal Antibody [clone: 523218]

Supplier: R & D Systems
MAB20471
220019-545EA 594.37 USD
220019-545
Anti-LY75 Mouse Monoclonal Antibody [clone: 523218]
Antibodies
Type: Primary
Antigen: LY75
Clonality: Monoclonal
Clone: 523218
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR