Anti-CD68 Mouse Monoclonal Antibody [clone: 298807]

Supplier: R & D Systems
MAB20401
220019-542EA 647.88 USD
220019-542
Anti-CD68 Mouse Monoclonal Antibody [clone: 298807]
Antibodies
Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 298807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR