Anti-CD68 Mouse Monoclonal Antibody [clone: 298813]

Supplier: R & D Systems
MAB2040
220019-541EA 600.15 USD
220019-541
Anti-CD68 Mouse Monoclonal Antibody [clone: 298813]
Antibodies
Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 298813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR