Anti-SPN Mouse Monoclonal Antibody [clone: 290111]

Supplier: R & D Systems
MAB2038
220019-539EA 594.37 USD
220019-539
Anti-SPN Mouse Monoclonal Antibody [clone: 290111]
Antibodies
Type: Primary
Antigen: SPN
Clonality: Monoclonal
Clone: 290111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR