Anti-BOC Mouse Monoclonal Antibody [clone: 273729]

Supplier: R & D Systems
MAB20361
220019-537EA 577.02 USD
220019-537
Anti-BOC Mouse Monoclonal Antibody [clone: 273729]
Antibodies
Type: Primary
Antigen: BOC
Clonality: Monoclonal
Clone: 273729
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR