Anti-NRCAM Mouse Monoclonal Antibody [clone: 297909]

Supplier: R & D Systems
MAB20341
220019-535EA 772.25 USD
220019-535
Anti-NRCAM Mouse Monoclonal Antibody [clone: 297909]
Antibodies
Type: Primary
Antigen: NRCAM
Clonality: Monoclonal
Clone: 297909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR