Anti-NRCAM Mouse Monoclonal Antibody [clone: 297906]

Supplier: R & D Systems
MAB2034
220019-534EA 633.42 USD
220019-534
Anti-NRCAM Mouse Monoclonal Antibody [clone: 297906]
Antibodies
Type: Primary
Antigen: NRCAM
Clonality: Monoclonal
Clone: 297906
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR