Anti-DDX4 Mouse Monoclonal Antibody [clone: 246904]

Supplier: R & D Systems
MAB2030
220019-533EA 772.25 USD
220019-533
Anti-DDX4 Mouse Monoclonal Antibody [clone: 246904]
Antibodies
Type: Primary
Antigen: DDX4
Clonality: Monoclonal
Clone: 246904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR