Anti-SMAD7 Mouse Monoclonal Antibody [clone: 293039]

Supplier: R & D Systems
MAB2029
220019-531EA 655.11 USD
220019-531
Anti-SMAD7 Mouse Monoclonal Antibody [clone: 293039]
Antibodies
Type: Primary
Antigen: SMAD7
Clonality: Monoclonal
Clone: 293039
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR