Anti-PROM2 Mouse Monoclonal Antibody [clone: 244029]

Supplier: R & D Systems
MAB2024
220019-529EA 633.42 USD
220019-529
Anti-PROM2 Mouse Monoclonal Antibody [clone: 244029]
Antibodies
Type: Primary
Antigen: PROM2
Clonality: Monoclonal
Clone: 244029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR