Anti-OV6 Mouse Monoclonal Antibody [clone: OV-6]

Supplier: R & D Systems
MAB2020
220019-528EA 647.88 USD
220019-528
Anti-OV6 Mouse Monoclonal Antibody [clone: OV-6]
Antibodies
Type: Primary
Antigen: OV6
Clonality: Monoclonal
Clone: OV-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR