Anti-SOX2 Mouse Monoclonal Antibody [clone: 245610]

Supplier: R & D Systems
MAB2018
220019-526EA 655.11 USD
220019-526
Anti-SOX2 Mouse Monoclonal Antibody [clone: 245610]
Antibodies
Simple Western

Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone: 245610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR