Anti-LILRB1 Mouse Monoclonal Antibody [clone: 292319]

Supplier: R & D Systems
MAB20172
220019-525EA 655.11 USD
220019-525
Anti-LILRB1 Mouse Monoclonal Antibody [clone: 292319]
Antibodies
Type: Primary
Antigen: LILRB1
Clonality: Monoclonal
Clone: 292319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR