Anti-LILRB1 Mouse Monoclonal Antibody [clone: 292305]

Supplier: R & D Systems
MAB20171
220019-524EA 647.88 USD
220019-524
Anti-LILRB1 Mouse Monoclonal Antibody [clone: 292305]
Antibodies
Type: Primary
Antigen: LILRB1
Clonality: Monoclonal
Clone: 292305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR