Anti-LILRB1 Mouse Monoclonal Antibody [clone: 292303]

Supplier: R & D Systems
MAB2017
220019-523EA 594.37 USD
220019-523
Anti-LILRB1 Mouse Monoclonal Antibody [clone: 292303]
Antibodies
Type: Primary
Antigen: LILRB1
Clonality: Monoclonal
Clone: 292303
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR