Anti-KIR2DL3 Mouse Monoclonal Antibody [clone: 180701]

Supplier: R & D Systems
MAB2014
220019-521EA 772.25 USD
220019-521
Anti-KIR2DL3 Mouse Monoclonal Antibody [clone: 180701]
Antibodies
Type: Primary
Antigen: KIR2DL3
Clonality: Monoclonal
Clone: 180701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR