Anti-RGMA Mouse Monoclonal Antibody [clone: 292012]

Supplier: R & D Systems
MAB2010
220019-520EA 561.11 USD
220019-520
Anti-RGMA Mouse Monoclonal Antibody [clone: 292012]
Antibodies
Type: Primary
Antigen: RGMA
Clonality: Monoclonal
Clone: 292012
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR