Anti-CD55 Mouse Monoclonal Antibody [clone: 278810]

Supplier: R & D Systems
MAB20091
220019-518EA 594.37 USD
220019-518
Anti-CD55 Mouse Monoclonal Antibody [clone: 278810]
Antibodies
Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 278810
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR