Anti-CD55 Mouse Monoclonal Antibody [clone: 278803]

Supplier: R & D Systems
MAB2009
220019-517EA 779.48 USD
220019-517
Anti-CD55 Mouse Monoclonal Antibody [clone: 278803]
Antibodies
Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 278803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR