Anti-KLK6 Mouse Monoclonal Antibody [clone: 265508]

Supplier: R & D Systems
MAB2008
220019-516EA 594.37 USD
220019-516
Anti-KLK6 Mouse Monoclonal Antibody [clone: 265508]
Antibodies
Type: Primary
Antigen: KLK6
Clonality: Monoclonal
Clone: 265508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR