Anti-RUNX2 Rat Monoclonal Antibody [clone: 232902]

Supplier: R & D Systems
MAB2006
220019-515EA 594.37 USD
220019-515
Anti-RUNX2 Rat Monoclonal Antibody [clone: 232902]
Antibodies
Type: Primary
Antigen: RUNX2
Clonality: Monoclonal
Clone: 232902
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR