Anti-CD46 Mouse Monoclonal Antibody [clone: 344519]

Supplier: R & D Systems
MAB2005
220019-514EA 647.88 USD
220019-514
Anti-CD46 Mouse Monoclonal Antibody [clone: 344519]
Antibodies
Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 344519
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR