Anti-OSCAR Mouse Monoclonal Antibody [clone: 259501]

Supplier: R & D Systems
MAB2004
220019-513EA 772.25 USD
220019-513
Anti-OSCAR Mouse Monoclonal Antibody [clone: 259501]
Antibodies
Type: Primary
Antigen: OSCAR
Clonality: Monoclonal
Clone: 259501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR