Anti-ROR1 Rat Monoclonal Antibody [clone: 291608]

Supplier: R & D Systems
MAB2000
220019-511EA 594.37 USD
220019-511
Anti-ROR1 Rat Monoclonal Antibody [clone: 291608]
Antibodies
Type: Primary
Antigen: ROR1
Clonality: Monoclonal
Clone: 291608
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR