Anti-ESM1 Rat Monoclonal Antibody [clone: 220008]

Supplier: R & D Systems
MAB1999
220019-509EA 772.25 USD
220019-509
Anti-ESM1 Rat Monoclonal Antibody [clone: 220008]
Antibodies
Type: Primary
Antigen: ESM1
Clonality: Monoclonal
Clone: 220008
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR