Anti-REG3B Mouse Monoclonal Antibody [clone: 293523]

Supplier: R & D Systems
MAB1996
220019-508EA 594.37 USD
220019-508
Anti-REG3B Mouse Monoclonal Antibody [clone: 293523]
Antibodies
Type: Primary
Antigen: REG3B
Clonality: Monoclonal
Clone: 293523
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR