Anti-PRSS27 Rat Monoclonal Antibody [clone: 246406]

Supplier: R & D Systems
MAB19891
102885-422EA 763.57 USD
102885-422
Anti-PRSS27 Rat Monoclonal Antibody [clone: 246406]
Antibodies
Type: Primary
Antigen: PRSS27
Clonality: Monoclonal
Clone: 246406
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR